Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000293.2 | NP_000284.1 | 17 | 1680 | ||
NM_001031835.2 | NP_001027005.1 | 18 | 1792 | |||
XM_005255983.4 | XP_005256040.1 | 17 | 1643 | |||
XM_005255984.4 | XP_005256041.1 | 18 | 1943 | |||
XM_017023282.1 | XP_016878771.1 | 11 | 937 | |||
XM_017023283.1 | XP_016878772.1 | 19 | 1848 | |||
XM_017023284.1 | XP_016878773.1 | 18 | 1729 | |||
GenBank mRNA | BC033657.1 | - | 18 | 1771 | 3913693 | |
CR936710.1 | - | 18 | 1824 | |||
X84908.1 | - | 17 | 1652 |