Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008534.1 | NP_001008534.1 | 2, 3 | 317 | ||
NM_001008535.1 | NP_001008535.1 | 2, 3 | 317 | |||
NM_178813.5 | NP_848928.1 | 2, 3 | 317 | |||
GenBank mRNA | AF514780.1 | - | 2, 3 | 264 | ||
AF514781.1 | - | 2, 3 | 264 | |||
AF514782.1 | - | 2, 3 | 264 | |||
BC066357.1 | - | 3, 4 | 454 | 5295545 | ||
BG208671.1 | - | 1, 2 | 241 |