Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001522.2 | NP_001513.2 | 2 | 554 | ||
GenBank mRNA | BC144241.1 | - | 1 | 349 | ||
L37378.1 | - | 2 | 577 |