Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320923.1 | NP_001307852.1 | 8, 9 | 1202 | ||
NM_001321852.1 | NP_001308781.1 | 7, 8 | 1205 | |||
NM_001321853.1 | NP_001308782.1 | 9, 10 | 1464 | |||
NM_001321854.1 | NP_001308783.1 | 8, 9 | 1380 | |||
NM_001321855.1 | NP_001308784.1 | 8, 9 | 1383 | |||
NM_001321856.1 | NP_001308785.1 | 7, 8 | 1118 | |||
NM_001321857.1 | NP_001308786.1 | 7, 8 | 1299 | |||
NM_002227.3 | NP_002218.2 | 7, 8 | 1299 | |||
XM_017001212.1 | XP_016856701.1 | 7, 8 | 1146 | |||
GenBank mRNA | AB209057.1 | - | 6, 7 | 1055 | ||
AB219242.1 | - | 5, 6 | 949 | |||
AK314257.1 | - | 8, 9 | 1204 | |||
BC009079.1 | - | 1234 | ||||
BC023604.2 | - | 990 | ||||
BC062431.1 | - | 7, 8 | 1234 | |||
BC111401.1 | - | 7, 8 | 1264 | |||
BC132729.1 | - | 6, 7 | 1017 | 40146751 | ||
HQ628631.1 | - | 5, 6 | 979 | |||
M64174.1 | - | 6, 7 | 1024 |