Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204182.1 | NP_001191111.1 | 7 | 1185 | ||
NM_001204183.1 | NP_001191112.1 | 7 | 1185 | |||
NM_002497.3 | NP_002488.1 | 7 | 1185 | |||
XM_005273147.1 | XP_005273204.1 | 7 | 1182 | |||
GenBank mRNA | AK223353.1 | - | 7 | 1146 | ||
AK225522.1 | - | 7 | 1146 | |||
AK303092.1 | - | 8 | 1048 | |||
AY045701.1 | - | 7 | 1116 | |||
AY863109.1 | - | 3 | 389 | |||
BC043502.1 | - | 7 | 1173 | 5541026 | ||
BT019729.1 | - | 7 | 1034 | |||
BU517064.1 | - | 2 | 261 | |||
DQ786247.1 | - | 1 | 65 | |||
DQ786281.1 | - | 1 | 22 | |||
U11050.1 | - | 7 | 1168 | |||
Z29066.1 | - | 7 | 1116 |