Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005990.3 | NP_005981.3 | 19 | 3433 | ||
XM_017009788.1 | XP_016865277.1 | 18 | 2987 | |||
GenBank mRNA | AB015718.1 | - | 19 | 3183 | ||
AF119894.1 | - | 1 | 130 | |||
AK022960.1 | - | 11 | 2006 | |||
AK092814.1 | - | 2 | 639 | |||
AL133081.1 | - | 4 | 694 | |||
BC014413.1 | - | 1 | 102 | |||
BC070077.1 | - | 19 | 3347 | 5267449 |