Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005990.3 | NP_005981.3 | 9 | 1629 | ||
XM_017009788.1 | XP_016865277.1 | 8 | 1183 | |||
GenBank mRNA | AB015718.1 | - | 9 | 1379 | ||
AK022960.1 | - | 1 | 202 | |||
AK307943.1 | - | 9 | 1163 | |||
AK313350.1 | - | 10 | 1384 | |||
BC070077.1 | - | 9 | 1543 | 5267449 |