Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318230.1 | NP_001305159.1 | 8 | 1175 | ||
NM_006285.2 | NP_006276.2 | 9 | 1258 | |||
GenBank mRNA | AF507907.1 | - | 8 | 671 | ||
AK298907.1 | - | 9 | 796 | |||
BC037561.1 | - | 9 | 853 | |||
BC038448.1 | - | 9 | 1232 | 5170888 | ||
BC067130.1 | - | 9 | 1269 | 6153652 | ||
D50863.1 | - | 9 | 1194 |