Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017021262.1 | XP_016876751.1 | 5, 6 | 710 | ||
XM_017021263.1 | XP_016876752.1 | 5, 6 | 436 | |||
XM_017021264.1 | XP_016876753.1 | 4, 5 | 508 | |||
GenBank mRNA | AF279372.1 | - | 6, 7 | 574 | ||
AK293515.1 | - | 6, 7 | 629 | |||
BC003622.2 | - | 7, 8 | 734 | 3010239 | ||
BC007428.2 | - | 7, 8 | 734 | 3010239 | ||
BC018192.1 | - | 7, 8 | 683 | 3867269 | ||
BC037305.2 | - | 5, 6 | 600 | |||
BX407816.2 | - | 7, 8 | 799 | |||
BX420122.2 | - | 7, 8 | 798 | |||
U51336.1 | - | 7, 8 | 627 |