Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPGT-TNNI3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112808.2 | NP_001106279.2 | 5 | 538 | ||
NM_001199327.1 | NP_001186256.2 | 5 | 538 | |||
GenBank mRNA | AB208941.1 | - | 5 | 487 | ||
BC032865.1 | - | 5 | 511 | 5263275 | ||
BX640903.1 | - | 5 | 523 |
Gene Symbol: TNNI3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015978.2 | NP_057062.1 | 3 | 233 | ||
GenBank mRNA | AF116826.1 | - | 3 | 219 | ||
AK307984.1 | - | 3 | 214 | |||
AY303691.1 | - | 3 | 198 | |||
BC113539.1 | - | 3 | 207 | 8322591 | ||
BC117262.1 | - | 3 | 207 | 40125813 | ||
BX502807.1 | - | 3 | 234 |