Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243736.1 | NP_001230665.1 | 7 | 550 | ||
NM_021970.3 | NP_068805.1 | 7 | 571 | |||
NR_024170.1 | 6 | 525 | ||||
GenBank mRNA | AF130115.1 | - | 6 | 371 | ||
AF201947.1 | - | 7 | 481 | |||
AK223311.1 | - | 7 | 462 | |||
AK311755.1 | - | 7 | 432 | |||
AL544981.3 | - | 7 | 380 | |||
BC026245.1 | - | 7 | 464 | 4808855 | ||
BC110402.1 | - | 6 | 391 | |||
BI562630.1 | - | 6 | 528 |