Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022755.5 | NP_073592.1 | 13 | 2117 | ||
XM_017015041.1 | XP_016870530.1 | 14 | 2187 | |||
GenBank mRNA | AF351200.1 | - | 1 | 339 | ||
AF351202.1 | - | 13 | 1923 | |||
AK023225.1 | - | 13 | 1907 | |||
AK024267.1 | - | 11 | 1695 | |||
AK096392.1 | - | 2 | 1435 | |||
AK123694.1 | - | 4 | 1763 | |||
BC026154.2 | - | 13 | 2119 | 4839268 |