Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHMK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184763.1 | NP_001171692.1 | 8 | 1597 | ||
NM_144624.2 | NP_653225.2 | 7 | 1849 | |||
NM_175866.4 | NP_787062.1 | 8 | 1938 | |||
GenBank mRNA | AJ536197.1 | - | 8 | 1887 | ||
AL834136.1 | - | 7 | 2323 | |||
BC014917.1 | - | 7 | 1427 |