Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000527.4 | NP_000518.1 | 7 | 1143 | ||
NM_001195798.1 | NP_001182727.1 | 7 | 1143 | |||
NM_001195799.1 | NP_001182728.1 | 6 | 1020 | |||
NM_001195800.1 | NP_001182729.1 | 5 | 639 | |||
NM_001195803.1 | NP_001182732.1 | 6 | 762 | |||
XM_011528010.1 | XP_011526312.1 | 7 | 1106 | |||
GenBank mRNA | AB209409.1 | - | 7 | 1027 | ||
AK295612.1 | - | 6 | 679 | |||
AK296312.1 | - | 6 | 661 | |||
AK299038.1 | - | 5 | 538 | |||
AK300313.1 | - | 6 | 919 | |||
AY114155.1 | - | 7 | 970 | |||
BC014514.1 | - | 7 | 1026 | 3542845 | ||
BT007361.1 | - | 7 | 956 | |||
BX648281.1 | - | 7 | 1211 | |||
S40543.1 | - | 1 | 25 |