Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000527.4 | NP_000518.1 | 18 | 3386 | ||
NM_001195798.1 | NP_001182727.1 | 18 | 3380 | |||
NM_001195799.1 | NP_001182728.1 | 17 | 3263 | |||
NM_001195800.1 | NP_001182729.1 | 16 | 2882 | |||
NM_001195803.1 | NP_001182732.1 | 16 | 2852 | |||
XM_011528010.1 | XP_011526312.1 | 17 | 3271 | |||
GenBank mRNA | BC014514.1 | - | 18 | 3269 | 3542845 | |
BX648281.1 | - | 18 | 3454 | |||
DA008286.1 | - | 1 | 502 |