Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025242.1 | NP_001020413.1 | 14 | 2582 | ||
NM_001025243.1 | NP_001020414.1 | 13 | 2435 | |||
NM_001569.3 | NP_001560.2 | 14 | 2672 | |||
XM_005274668.3 | XP_005274725.1 | 13 | 2604 | |||
XM_011531158.2 | XP_011529460.1 | 13 | 2360 | |||
GenBank mRNA | BC014963.1 | - | 13 | 2420 | 4906853 | |
BC054000.1 | - | 14 | 2614 | 6164719 | ||
L76191.1 | - | 14 | 2672 |