Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178844.3 | NP_849172.2 | 20 | 3611 | ||
NR_075083.2 | 21 | 3808 | ||||
XM_017023027.1 | XP_016878516.1 | 20 | 3641 | |||
XM_017023028.1 | XP_016878517.1 | 21 | 3815 | |||
XM_017023029.1 | XP_016878518.1 | 22 | 3896 | |||
XM_017023030.1 | XP_016878519.1 | 22 | 3957 | |||
XM_017023031.1 | XP_016878520.1 | 20 | 3702 | |||
XM_017023032.1 | XP_016878521.1 | 22 | 4658 | |||
XM_017023033.1 | XP_016878522.1 | 22 | 3835 | |||
XM_017023034.1 | XP_016878523.1 | 21 | 3754 | |||
XM_017023035.1 | XP_016878524.1 | 21 | 3783 | |||
XM_017023036.1 | XP_016878525.1 | 20 | 4342 | |||
XM_017023037.1 | XP_016878526.1 | 21 | 3536 | |||
XM_017023038.1 | XP_016878527.1 | 21 | 3509 | |||
GenBank mRNA | AK074109.1 | - | 16 | 1489 | ||
AK090431.1 | - | 21 | 3806 | |||
AY601811.1 | - | 20 | 3598 | |||
FJ889357.1 | - | 17 | 3187 |