Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006908.4 | NP_008839.2 | 6 | 2015 | ||
NM_018890.3 | NP_061485.1 | 7 | 2072 | |||
GenBank mRNA | AK054993.1 | - | 6 | 1990 | ||
BC050687.1 | - | 6 | 1962 | 6149377 | ||
D25274.1 | - | 1 | 906 |