Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016249.3 | NP_057333.1 | 3 | 1788 | ||
GenBank mRNA | AF116194.1 | - | 3 | 1771 | ||
AF151378.1 | - | 3 | 1769 | |||
AF196482.1 | - | 3 | 1769 | |||
BC005891.2 | - | 1 | 1519 | 3050294 | ||
BC013318.1 | - | 3 | 1792 | 4133159 | ||
BM548987.1 | - | 1 | 244 |