Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YTHDF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172828.1 | NP_001166299.1 | 3 | 1859 | ||
NM_001173128.1 | NP_001166599.1 | 5 | 1966 | |||
NM_016258.2 | NP_057342.2 | 4 | 2200 | |||
GenBank mRNA | AF155095.1 | - | 4 | 1854 | ||
AF192968.1 | - | 4 | 1854 | |||
AF432214.1 | - | 3 | 2383 | |||
AK074276.1 | - | 1 | 1275 | |||
AK292581.1 | - | 4 | 2002 | |||
AK303833.1 | - | 3 | 1859 | |||
BC002559.2 | - | 4 | 2065 | 3139250 |