Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278666.1 | NP_001265595.1 | 3 | 400 | ||
NM_001278667.1 | NP_001265596.1 | 3 | 572 | |||
NM_001278668.1 | NP_001265597.1 | 3 | 399 | |||
NM_016277.4 | NP_057361.3 | 3 | 869 | |||
NM_183227.2 | NP_899050.1 | 3 | 423 | |||
GenBank mRNA | AB025427.1 | - | 2 | 215 | ||
AB034244.1 | - | 3 | 368 | |||
AF161486.1 | - | 3 | 365 | |||
AF498951.1 | - | 2 | 215 | |||
AK313796.1 | - | 3 | 420 | |||
AU280196.1 | - | 3 | 399 | |||
AY585189.1 | - | 2 | 215 | |||
BC015021.1 | - | 3 | 835 | 3910708 | ||
CR749371.1 | - | 4 | 603 | |||
DA021294.1 | - | 3 | 423 |