Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290007.1 | NP_001276936.1 | 13 | 2011 | ||
NM_001290008.1 | NP_001276937.1 | 12 | 1924 | |||
NM_001290009.1 | NP_001276938.1 | 12 | 2590 | |||
NM_016293.3 | NP_057377.3 | 13 | 2020 | |||
XM_005268957.3 | XP_005269014.2 | 13 | 1993 | |||
XM_005268958.3 | XP_005269015.1 | 12 | 1939 | |||
XM_017019422.1 | XP_016874911.1 | 10 | 1774 | |||
GenBank mRNA | AB032698.1 | - | 13 | 1962 | ||
AF146531.1 | - | 13 | 1932 | |||
AK000783.1 | - | 12 | 1879 | |||
AK225911.1 | - | 12 | 2546 | |||
BC047686.1 | - | 13 | 1986 | 5722815 | ||
BG388638.1 | - | 3 | 409 | |||
BX281612.1 | - | 3 | 410 |