Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164372.1 | NP_001157844.1 | 8 | 1352 | ||
NM_001164373.1 | NP_001157845.1 | 8 | 1261 | |||
NM_016301.3 | NP_057385.3 | 8 | 1231 | |||
XM_005253896.4 | XP_005253953.3 | 9 | 1686 | |||
XM_017019394.1 | XP_016874883.1 | 8 | 1166 | |||
GenBank mRNA | AF117229.1 | - | 8 | 1181 | ||
AK223209.1 | - | 8 | 1203 | |||
BC008416.1 | - | 8 | 1206 | 4048678 | ||
BC031024.1 | - | 8 | 1207 | 4695834 | ||
BX537973.1 | - | 8 | 1242 |