Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303533.1 | NP_001290462.1 | 15 | 2725 | ||
NM_001303534.1 | NP_001290463.1 | 15 | 2820 | |||
NM_016339.4 | NP_057423.1 | 15 | 3005 | |||
XM_006721932.3 | XP_006721995.1 | 15 | 3499 | |||
XM_006721935.3 | XP_006721998.1 | 16 | 2776 | |||
XM_006721937.3 | XP_006722000.1 | 15 | 4601 | |||
XM_006721938.3 | XP_006722001.1 | 12 | 2392 | |||
GenBank mRNA | AF117946.1 | - | 15 | 2625 | ||
BC115374.1 | - | 13 | 2528 | 40028278 | ||
BC115375.1 | - | 13 | 2528 | 40028280 |