Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198976.2 | NP_945327.2 | 1, 2 | 173 | ||
GenBank mRNA | AF161479.1 | - | 1, 2 | 105 | ||
AJ238379.1 | - | 1, 2 | 107 | |||
AK001316.1 | - | 1, 2 | 81 | |||
AK023310.1 | - | 1, 2 | 98 | |||
AK023927.1 | - | 1, 2 | 98 | |||
AK293410.1 | - | 1, 2 | 125 | |||
AK304310.1 | - | 1, 2 | 125 | |||
BC014952.1 | - | 1, 2 | 109 | 4860894 | ||
BI459250.1 | - | 1, 2 | 134 | |||
BX647417.1 | - | 1, 2 | 108 | |||
HY015074.1 | - | 1, 2 | 174 |