Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286082.1 | NP_001273011.1 | 3 | 767 | ||
NM_001286084.1 | NP_001273013.1 | 3 | 782 | |||
NM_016404.2 | NP_057488.1 | 3 | 782 | |||
GenBank mRNA | AF110774.1 | - | 3 | 280 | ||
AF161501.1 | - | 3 | 259 | |||
AF229068.1 | - | 3 | 259 | |||
AK312050.1 | - | 3 | 224 | |||
BC017172.2 | - | 3 | 248 | 3163327 | ||
BC029482.2 | - | 3 | 236 | 4737961 | ||
BC105294.1 | - | 3 | 238 | 6725102 | ||
BE791577.1 | - | 3 | 277 | |||
CB054126.1 | - | 1 | 27 | |||
CN362692.1 | - | 3 | 230 |