Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016406.3 | NP_057490.2 | 1 | 361 | ||
XM_005245254.1 | XP_005245311.1 | 1 | 313 | |||
XM_017001450.1 | XP_016856939.1 | 2 | 206 | |||
XM_017001451.1 | XP_016856940.1 | 2 | 470 | |||
GenBank mRNA | AB154405.1 | - | 1 | 107 | ||
AF151884.1 | - | 1 | 328 | |||
AF161504.1 | - | 1 | 347 | |||
AK292776.1 | - | 1 | 313 | |||
BC005187.1 | - | 1 | 353 | 3961569 | ||
CF128121.1 | - | 1 | 360 |