Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK5RAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278167.1 | NP_001265096.1 | 2 | 329 | ||
NM_001278168.1 | NP_001265097.1 | 2 | 329 | |||
NM_016082.4 | NP_057166.4 | 2 | 181 | |||
NM_016408.3 | NP_057492.2 | 2 | 329 | |||
XM_011528858.1 | XP_011527160.1 | 2 | 284 | |||
XM_017027876.1 | XP_016883365.1 | 2 | 290 | |||
GenBank mRNA | AF152097.1 | - | 2 | 268 | ||
AF161516.1 | - | 2 | 280 | |||
AK023992.1 | - | 2 | 279 | |||
AK027449.1 | - | 2 | 129 | |||
AK222745.1 | - | 2 | 279 | |||
AK295342.1 | - | 2 | 279 | |||
AK309906.1 | - | 2 | 279 | |||
AK314532.1 | - | 2 | 279 | |||
AY462283.1 | - | 2 | 280 | |||
AY462284.1 | - | 2 | 280 | |||
BC001215.1 | - | 2 | 258 | 3535564 | ||
BC050706.1 | - | 2 | 272 | 4418659 | ||
BM834342.1 | - | 2 | 132 | |||
BQ072829.1 | - | 291 | ||||
DA028522.1 | - | 2 | 279 | |||
DA565688.1 | - | 2 | 131 | |||
DC427896.1 | - | 2 | 329 | |||
KU178644.1 | - | 1 | 167 | |||
KU178645.1 | - | 1 | 167 |