Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271880.1 | NP_001258809.1 | 4 | 457 | ||
NM_016438.3 | NP_057522.1 | 3 | 541 | |||
NR_073504.1 | 2 | 406 | ||||
XM_006721946.2 | XP_006722009.1 | 4 | 435 | |||
XM_006721947.3 | XP_006722010.1 | 4 | 845 | |||
XM_006721948.3 | XP_006722011.1 | 4 | 539 | |||
XM_011524891.2 | XP_011523193.1 | 4 | 939 | |||
XM_017024742.1 | XP_016880231.1 | 4 | 885 | |||
XM_017024743.1 | XP_016880232.1 | 4 | 977 | |||
GenBank mRNA | AB038021.1 | - | 3 | 368 | ||
AB038022.1 | - | 3 | 368 | |||
AB095269.1 | - | 1 | 91 | |||
BC020667.1 | - | 3 | 532 | 4724613 | ||
BG573492.1 | - | 3 | 533 | |||
BM503772.1 | - | 3 | 544 | |||
BM876435.1 | - | 2 | 382 |