Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016445.2 | NP_057529.1 | 2 | 215 | ||
XM_011536644.2 | XP_011534946.1 | 2 | 189 | |||
GenBank mRNA | AF228603.1 | - | 2 | 126 | ||
BC001226.1 | - | 2 | 159 | 3457876 | ||
BC008056.2 | - | 2 | 69 | |||
CR976852.1 | - | 2 | 219 |