Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCKIPSD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016453.3 | NP_057537.1 | 6 | 1379 | ||
NM_184231.2 | NP_909119.1 | 6 | 1358 | |||
XM_017006594.1 | XP_016862083.1 | 6 | 1373 | |||
XM_017006595.1 | XP_016862084.1 | 6 | 1366 | |||
XM_017006596.1 | XP_016862085.1 | 6 | 1393 | |||
XM_017006597.1 | XP_016862086.1 | 6 | 1137 | |||
XM_017006598.1 | XP_016862087.1 | 6 | 1363 | |||
XM_017006599.1 | XP_016862088.1 | 6 | 1399 | |||
XM_017006600.1 | XP_016862089.1 | 6 | 1362 | |||
GenBank mRNA | AF178432.1 | - | 6 | 1347 | ||
AF303581.1 | - | 6 | 1318 | |||
AJ242655.2 | - | 6 | 1347 | |||
AY453794.1 | - | 6 | 1326 | |||
BC016052.1 | - | 6 | 1242 | 4862348 | ||
BC026280.1 | - | 6 | 1309 | 4730420 |