Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000078.2 | NP_000069.2 | 10, 11 | 1031 | ||
NM_001286085.1 | NP_001273014.1 | 9, 10 | 851 | |||
GenBank mRNA | AK291586.1 | - | 10, 11 | 991 | ||
AK297699.1 | - | 9, 10 | 855 | |||
BC025739.1 | - | 10, 11 | 990 | 5223614 | ||
BT020064.1 | - | 9, 10 | 794 | |||
KU177972.1 | - | 10, 11 | 974 | |||
KU177973.1 | - | 9, 10 | 794 | |||
M30185.1 | - | 10, 11 | 1104 | |||
M83573.1 | - | 9, 10 | 771 |