Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHISA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272065.1 | NP_001258994.1 | 6 | 1195 | ||
NM_001272066.1 | NP_001258995.1 | 6 | 1182 | |||
NM_001272067.1 | NP_001258996.1 | 6 | 1377 | |||
NM_001272068.1 | NP_001258997.1 | 7 | 1414 | |||
NM_001272082.1 | NP_001259011.1 | 4 | 1071 | |||
NM_001272083.1 | NP_001259012.1 | 3 | 1245 | |||
NM_016479.4 | NP_057563.3 | 6 | 1216 | |||
XM_005265205.1 | XP_005265262.1 | 7 | 1464 | |||
XM_006713188.2 | XP_006713251.1 | 6 | 1117 | |||
XM_011533796.1 | XP_011532098.1 | 7 | 1206 | |||
GenBank mRNA | AB097007.1 | - | 4 | 835 | ||
AF151051.1 | - | 5 | 839 | |||
AF520698.1 | - | 6 | 1195 | |||
AK027728.1 | - | 438 | ||||
AK056328.1 | - | 6 | 1358 | |||
AK075441.1 | - | 6 | 1131 | |||
AK124652.1 | - | 7 | 1414 | |||
AK298799.1 | - | 4 | 827 | |||
AK298842.1 | - | 4 | 846 | |||
AK310083.1 | - | 2 | 1193 | |||
AL117413.1 | - | 3 | 1016 | |||
AL832568.1 | - | 4 | 878 | |||
BC001463.2 | - | 4 | 886 | 3138644 | ||
BC020279.2 | - | 4 | 836 |