Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016484.4 | NP_057568.1 | 4, 5 | 338 | ||
XM_011530973.2 | XP_011529275.1 | 5, 6 | 502 | |||
XM_017029568.1 | XP_016885057.1 | 4, 5 | 669 | |||
GenBank mRNA | AF151061.1 | - | 4, 5 | 308 | ||
AK024746.1 | - | 4, 5 | 306 | |||
AK311539.1 | - | 3, 4 | 200 | |||
AY358829.1 | - | 4, 5 | 544 | |||
BC012996.2 | - | 4, 5 | 277 | 3530647 | ||
BC089433.1 | - | 3, 4 | 235 | |||
BG708698.1 | - | 4, 5 | 342 | |||
BX537725.1 | - | 4, 5 | 412 | |||
CR457149.1 | - | 3, 4 | 236 |