Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM178B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122646.2 | NP_001116118.2 | 17 | 2574 | ||
NM_001172667.1 | NP_001166138.1 | 5 | 775 | |||
NM_016490.4 | NP_057574.2 | 5 | 893 | |||
XM_011511294.2 | XP_011509596.1 | 16 | 2502 | |||
XM_017004263.1 | XP_016859752.1 | 18 | 2739 | |||
XM_017004264.1 | XP_016859753.1 | 18 | 2700 | |||
XM_017004266.1 | XP_016859755.1 | 8 | 1147 | |||
XM_017004267.1 | XP_016859756.1 | 5 | 752 | |||
XM_017004268.1 | XP_016859757.1 | 5 | 778 | |||
GenBank mRNA | AF151068.1 | - | 5 | 733 | ||
AJ710728.1 | - | 2 | 295 | |||
AK122705.1 | - | 11 | 1453 | |||
BC038583.1 | - | 5 | 854 | |||
BC039488.1 | - | 15 | 1841 | |||
BC051379.1 | - | 5 | 893 |