Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003704.2 | NP_001003704.1 | 3 | 681 | ||
NM_016498.4 | NP_057582.2 | 4 | 914 | |||
GenBank mRNA | AF060924.1 | - | 4 | 825 | ||
AF151076.1 | - | 4 | 875 | |||
AK026104.1 | - | 4 | 782 | |||
BC001608.1 | - | 4 | 740 | 3542567 | ||
BC009300.2 | - | 4 | 772 | 4126237 | ||
BC014446.1 | - | 4 | 740 | 4855192 | ||
BC030989.2 | - | 4 | 801 | 4077757 | ||
BC038831.2 | - | 4 | 805 | 3908299 | ||
BC046132.1 | - | 4 | 704 | 4446034 | ||
BQ187590.1 | - | 3 | 463 |