Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282568.1 | NP_001269497.1 | 2, 3 | 361 | ||
NM_001282571.1 | NP_001269500.1 | 2, 3 | 223 | |||
NM_001282572.1 | NP_001269501.1 | 2, 3 | 223 | |||
NM_001282574.1 | NP_001269503.1 | 2, 3 | 223 | |||
NM_016505.3 | NP_057589.2 | 2, 3 | 223 | |||
XM_011541567.2 | XP_011539869.1 | 2, 3 | 239 | |||
GenBank mRNA | AF151077.1 | - | 2, 3 | 152 | ||
AF151085.1 | - | 2, 3 | 181 | |||
AK001929.1 | - | 2, 3 | 173 | |||
AK022506.1 | - | 2, 3 | 210 | |||
AK024049.1 | - | 2, 3 | 199 | |||
BC000685.2 | - | 2, 3 | 261 | |||
BC007446.2 | - | 2, 3 | 159 | 3009823 | ||
BC050609.1 | - | 2, 3 | 163 | 6527025 | ||
BF309574.1 | - | 2, 3 | 174 | |||
BP309837.1 | - | 2, 3 | 164 | |||
BP371153.1 | - | 2, 3 | 159 | |||
CN423139.1 | - | 2, 3 | 160 | |||
DC366453.1 | - | 2, 3 | 361 |