Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016523.2 | NP_057607.1 | 3 | 264 | ||
XM_017019414.1 | XP_016874903.1 | 3 | 383 | |||
XM_017019415.1 | XP_016874904.1 | 3 | 455 | |||
GenBank mRNA | AF175206.1 | - | 3 | 264 | ||
AJ305370.1 | - | 3 | 247 | |||
BC096734.1 | - | 3 | 224 | |||
BC098166.1 | - | 3 | 224 | 40015535 | ||
BC098257.1 | - | 3 | 224 | 40015534 | ||
BC098354.1 | - | 3 | 224 | 40015531 |