Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000214.2 | NP_000205.1 | 26 | 5502 | ||
XM_017027707.1 | XP_016883196.1 | 25 | 5607 | |||
GenBank mRNA | AF003837.1 | - | 26 | 5444 | ||
AF028593.1 | - | 26 | 5248 | |||
BC098393.1 | - | 26 | 5404 | 30528888 | ||
U73936.1 | - | 26 | 5398 | |||
U77720.1 | - | 26 | 4948 | |||
U77914.1 | - | 10 | 1402 |