Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113546.1 | NP_001107018.1 | 4 | 629 | ||
NM_001113547.1 | NP_001107019.1 | 1 | 510 | |||
NM_016357.4 | NP_057441.1 | 4 | 629 | |||
XM_011538455.2 | XP_011536757.1 | 4 | 689 | |||
XM_017019430.1 | XP_016874919.1 | 4 | 543 | |||
GenBank mRNA | AB209512.1 | - | 4 | 565 | ||
AF198454.1 | - | 4 | 556 | |||
AF198455.1 | - | 1 | 448 | |||
AK000335.1 | - | 1 | 153 | |||
AK000372.1 | - | 4 | 539 | |||
AK222971.1 | - | 4 | 552 | |||
AK297766.1 | - | 2 | 225 | |||
AK314447.1 | - | 4 | 541 | |||
AL136911.1 | - | 4 | 548 | |||
BC001247.1 | - | 1 | 161 | |||
BC110815.1 | - | 4 | 566 | 5115521 | ||
BC136763.1 | - | 4 | 515 | 9020753 | ||
BC144402.1 | - | 4 | 515 | |||
BC144403.1 | - | 4 | 515 | |||
DA738195.1 | - | 1 | 510 |
Gene Symbol: C5orf24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK130086.1 | - | 1677 |