Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUZP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318840.1 | NP_001305769.1 | 2 | 222 | ||
NM_016383.4 | NP_057467.1 | 2 | 222 | |||
GenBank mRNA | AF124430.1 | - | 2 | 192 | ||
AK093369.1 | - | 2 | 213 | |||
AK314765.1 | - | 2 | 211 | |||
BC128134.1 | - | 2 | 184 | 40112435 | ||
BC128135.1 | - | 2 | 184 | |||
DB035084.1 | - | 2 | 223 | |||
DB041564.1 | - | 2 | 214 |