Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256539.3 | NP_001243468.2 | 3 | 639 | ||
NM_001256540.3 | NP_001243469.2 | 3 | 639 | |||
NM_001291305.2 | NP_001278234.1 | 3 | 673 | |||
NM_001291307.2 | NP_001278236.1 | 3 | 639 | |||
NM_001317975.1 | NP_001304904.1 | 2 | 728 | |||
NM_016391.7 | NP_057475.2 | 3 | 639 | |||
NR_134316.1 | 2 | 745 | ||||
NR_134318.1 | 3 | 639 | ||||
GenBank mRNA | AF151019.1 | - | 3 | 635 | ||
AF151875.1 | - | 4 | 707 | |||
AF161460.1 | - | 3 | 313 | |||
AK300883.1 | - | 3 | 318 | |||
AK303281.1 | - | 3 | 310 | |||
AW837047.1 | - | 2 | 394 | |||
AY561704.1 | - | 3 | 251 | |||
BC019331.1 | - | 3 | 474 | 3534019 | ||
BC032424.2 | - | 3 | 282 | 5416724 | ||
BC040106.1 | - | 3 | 299 | 6066208 | ||
BP234677.1 | - | 2 | 399 | |||
BU594763.1 | - | 3 | 313 | |||
CB122385.1 | - | 3 | 299 | |||
CR457169.1 | - | 3 | 251 | |||
HY064071.1 | - | 3 | 322 |