Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERF2-C15ORF63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037673.1 | 3 | 788 | |||
GenBank mRNA | AF370428.1 | - | 2 | 4281 | ||
AK000438.1 | - | 3 | 410 | |||
AK094716.1 | - | 2 | 2113 | |||
AK095876.1 | - | 4 | 1786 |
Gene Symbol: HYPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199885.1 | NP_001186814.1 | 1 | 345 | ||
NM_016400.3 | NP_057484.3 | 1 | 345 | |||
GenBank mRNA | AF049613.1 | - | 1 | 142 | ||
AF161485.1 | - | 1 | 166 | |||
AK022435.1 | - | 1 | 167 | |||
BC019262.1 | - | 1 | 343 | 3504222 | ||
BQ478474.1 | - | 1 | 143 |