Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016464.4 | NP_057548.1 | 5 | 1032 | ||
NR_028473.1 | 6 | 915 | ||||
XM_006718585.3 | XP_006718648.1 | 4 | 1270 | |||
XM_006718586.2 | XP_006718649.1 | 5 | 669 | |||
XM_006718588.3 | XP_006718651.1 | 5 | 527 | |||
GenBank mRNA | AF151030.1 | - | 5 | 649 | ||
AF151032.1 | - | 5 | 646 | |||
AK027724.1 | - | 5 | 1181 | |||
AL832879.1 | - | 3 | 3185 | |||
BC005201.1 | - | 5 | 647 | 3684390 | ||
CB956757.1 | - | 6 | 587 |