Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128150.1 | NP_001121622.1 | 3 | 349 | ||
NM_016467.4 | NP_057551.1 | 4 | 459 | |||
XM_005246970.2 | XP_005247027.1 | 3 | 378 | |||
XM_011512199.2 | XP_011510501.1 | 3 | 341 | |||
XM_011512200.2 | XP_011510502.1 | 2 | 1956 | |||
XM_017005347.1 | XP_016860836.1 | 4 | 498 | |||
XM_017005348.1 | XP_016860837.1 | 3 | 330 | |||
GenBank mRNA | AB064959.1 | - | 2 | 918 | ||
AB064960.1 | - | 3 | 357 | |||
AF151036.1 | - | 3 | 297 | |||
AF395704.1 | - | 4 | 414 | |||
AK074756.1 | - | 3 | 348 | |||
AK075160.1 | - | 2 | 350 | |||
AK313533.1 | - | 4 | 424 | |||
BC005200.1 | - | 3 | 316 | 3929844 | ||
CR936635.1 | - | 2 | 1026 | |||
DB122266.1 | - | 3 | 349 |