Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NT5C3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002009.2 | NP_001002009.1 | 3 | 375 | ||
NM_001002010.2 | NP_001002010.1 | 2 | 320 | |||
NM_001166118.2 | NP_001159590.1 | 4 | 525 | |||
NM_016489.12 | NP_057573.2 | 3 | 439 | |||
XM_011515409.2 | XP_011513711.1 | 4 | 918 | |||
GenBank mRNA | AF151067.1 | - | 2 | 161 | ||
AF312735.1 | - | 2 | 256 | |||
AK290118.1 | - | 2 | 277 | |||
AK314109.1 | - | 2 | 319 | |||
AL136716.1 | - | 2 | 285 | |||
BC013292.2 | - | 2 | 203 | |||
BC015856.1 | - | 3 | 184 | 4669009 | ||
BC066914.1 | - | 3 | 300 | 5264738 | ||
BC071652.1 | - | 2 | 302 | 3892369 | ||
BF790086.1 | - | 3 | 185 | |||
CB127255.1 | - | 2 | 320 | |||
CB127877.1 | - | 4 | 264 | |||
CR533518.1 | - | 2 | 243 | |||
DA975244.1 | - | 3 | 439 | |||
DB243020.1 | - | 4 | 269 |