Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000662.7 | NP_000653.3 | 3 | 951 | ||
NM_001160170.3 | NP_001153642.1 | 5 | 1485 | |||
NM_001160171.3 | NP_001153643.1 | 4 | 1335 | |||
NM_001160172.3 | NP_001153644.1 | 5 | 1256 | |||
NM_001160173.3 | NP_001153645.1 | 4 | 1101 | |||
NM_001160174.2 | NP_001153646.1 | 1 | 1168 | |||
NM_001160175.3 | NP_001153647.1 | 5 | 1219 | |||
NM_001160176.3 | NP_001153648.1 | 4 | 1069 | |||
NM_001160179.2 | NP_001153651.1 | 5 | 1155 | |||
NM_001291962.1 | NP_001278891.1 | 6 | 1273 | |||
XM_006716410.3 | XP_006716473.1 | 3 | 1528 | |||
XM_011544687.1 | XP_011542989.1 | 6 | 1614 | |||
XM_011544688.1 | XP_011542990.1 | 5 | 1464 | |||
XM_011544689.2 | XP_011542991.1 | 2 | 1313 | |||
XM_017013947.1 | XP_016869436.1 | 8 | 1850 | |||
GenBank mRNA | AJ581142.1 | - | 1 | 45 | ||
AJ581143.1 | - | 1 | 45 | |||
AK290912.1 | - | 3 | 1426 | |||
BC013732.1 | - | 4 | 1308 | |||
BC047666.1 | - | 3 | 930 | 6059458 | ||
BX647521.1 | - | 4 | 1044 | |||
D90041.1 | - | 1 | 934 |