Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NTM SIRNA Assays ›
Gene Symbol:
NTM
Gene Name:
neurotrimin
Gene Aliases:
HNT, IGLON2, NTRI
Chromosome Location:
Chr.11: 131370326 - 132336822 on Build GRCh38
UniGene ID:
Hs.504352
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
24499
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NTM
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001048209.1
NP_001041674.1
4
787
NM_001144058.1
NP_001137530.1
3
1120
NM_001144059.1
NP_001137531.1
3
1120
NM_016522.2
NP_057606.1
3
1120
XM_005271580.3
XP_005271637.1
4
937
XM_005271581.3
XP_005271638.1
3
890
XM_005271582.3
XP_005271639.1
4
937
XM_006718855.3
XP_006718918.1
5
1044
XM_006718857.3
XP_006718920.1
4
964
XM_011542861.2
XP_011541163.1
3
890
XM_017017851.1
XP_016873340.1
3
890
XM_017017852.1
XP_016873341.1
4
937
XM_017017853.1
XP_016873342.1
5
1044
XM_017017854.1
XP_016873343.1
3
890
XM_017017855.1
XP_016873344.1
4
937
XM_017017856.1
XP_016873345.1
5
650
XM_017017857.1
XP_016873346.1
5
941
XM_017017858.1
XP_016873347.1
5
934
XM_017017859.1
XP_016873348.1
5
650
XM_017017860.1
XP_016873349.1
4
967
XM_017017861.1
XP_016873350.1
5
1044
XM_017017862.1
XP_016873351.1
5
931
XM_017017863.1
XP_016873352.1
4
967
XM_017017864.1
XP_016873353.1
4
935
XM_017017865.1
XP_016873354.1
4
937
XM_017017866.1
XP_016873355.1
4
964
XM_017017867.1
XP_016873356.1
2
144
GenBank mRNA
AF126426.1
-
3
720
AK293448.1
-
4
755
AK293487.1
-
5
736
AK294137.1
-
4
666
AK315964.1
-
4
782
AY358331.1
-
4
589
BC050716.1
-
3
470
6166839
BE263639.1
-
2
186
EF043567.1
-
3
476
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
neuron recognition
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0