Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADORA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000675.5 | NP_000666.2 | 3 | 2146 | ||
NM_001278497.1 | NP_001265426.1 | 4 | 2317 | |||
NM_001278498.1 | NP_001265427.1 | 3 | 2211 | |||
NM_001278499.1 | NP_001265428.1 | 3 | 2126 | |||
NM_001278500.1 | NP_001265429.1 | 3 | 1993 | |||
NR_103543.1 | 2 | 1540 | ||||
NR_103544.1 | 2 | 1520 | ||||
GenBank mRNA | AK123058.1 | - | 4 | 2056 | ||
BC013780.1 | - | 3 | 2073 | 4385637 | ||
M97370.1 | - | 2 | 1948 |
Gene Symbol: SPECC1L-ADORA2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103546.1 | 20 | 5866 | |||
GenBank mRNA | AB209022.1 | - | 20 | 5783 |
Set Membership: |
![]() |