Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016355.3 | NP_057439.2 | 10 | 1116 | ||
NM_201224.1 | NP_957518.1 | 9 | 969 | |||
GenBank mRNA | AF078843.1 | - | 9 | 926 | ||
AF190165.1 | - | 9 | 1629 | |||
AJ276704.1 | - | 10 | 1094 | |||
AK054574.1 | - | 10 | 1101 | |||
AK222940.1 | - | 10 | 1075 | |||
AK225144.1 | - | 10 | 1086 | |||
AL136666.1 | - | 10 | 1120 | |||
AL833469.1 | - | 7 | 1989 | |||
BC009379.2 | - | 10 | 1079 | |||
BC068009.1 | - | 10 | 1098 | 6142837 |